Assay Details
| Gene Symbol: UBAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283377.1 | NP_001270306.1 | 4 - 5 | 472 | 90 | |
| XM_005586160.2 | XP_005586217.1 | 4 - 5 | 387 | 90 | ||
| GenBank mRNA | AB168287.1 | - | 4 - 5 | 472 | 90 | |
More Information
Set Membership: |
|

UBAC2
Genomic Map