Assay Details
| Gene Symbol: SRI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287327.1 | NP_001274256.1 | 2 - 3 | 163 | 74 | |
| XM_005550298.2 | XP_005550355.1 | 2 - 3 | 174 | 74 | ||
| XM_015447317.1 | XP_015302803.1 | 2 - 3 | 207 | 74 | ||
| GenBank mRNA | AB173436.1 | - | 2 - 3 | 163 | 74 | |
More Information
Set Membership: |
|

SRI
Genomic Map