Assay Details
| Gene Symbol: IRAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547799.2 | XP_005547856.1 | 1 - 2 | 179 | 57 | |
| XM_005547800.2 | XP_005547857.1 | 1 - 2 | 161 | 57 | ||
| XM_015446262.1 | XP_015301748.1 | 1 - 2 | 162 | 57 | ||
More Information
Set Membership: |
|

IRAK2
Genomic Map