Assay Details
| Gene Symbol: ZC3HC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550757.1 | XP_005550814.1 | 4 - 5 | 573 | 92 | |
| GenBank mRNA | AB174426.1 | - | 4 - 5 | 505 | 92 | |
More Information
Set Membership: |
|

ZC3HC1
Genomic Map