Assay Details
| Gene Symbol: PRDM11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578110.2 | XP_005578167.1 | 5 - 6 | 1692 | 79 | |
| XM_005578111.2 | XP_005578168.1 | 5 - 6 | 910 | 79 | ||
| XM_005578114.2 | XP_005578171.1 | 5 - 6 | 1691 | 79 | ||
| XM_005578116.2 | XP_005578173.1 | 5 - 6 | 1687 | 79 | ||
| XM_015434867.1 | XP_015290353.1 | 2 - 3 | 5221 | 79 | ||
| GenBank mRNA | AB170322.1 | - | 689 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015434867.1 transcripts.
Set Membership: |
|

PRDM11
Genomic Map