Assay Details
| Gene Symbol: LOC102133131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570067.2 | XP_005570124.2 | 5 - 6 | 567 | 69 | |
| XM_005570068.2 | XP_005570125.1 | 5 - 6 | 567 | 69 | ||
More Information
Set Membership: |
|

LOC102133131
Genomic Map