Assay Details
| Gene Symbol: MAP7D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594697.2 | XP_005594754.1 | 3 - 4 | 459 | 113 | |
| XM_015443905.1 | XP_015299391.1 | 3 - 4 | 459 | 113 | ||
| XM_015443906.1 | XP_015299392.1 | 3 - 4 | 459 | 113 | ||
| XM_015443907.1 | XP_015299393.1 | 3 - 4 | 459 | 113 | ||
| XM_015443908.1 | XP_015299394.1 | 3 - 4 | 459 | 113 | ||
| GenBank mRNA | AB170232.1 | - | 3 - 4 | 265 | 113 | |
More Information
Set Membership: |
|

MAP7D3
Genomic Map