Assay Details
| Gene Symbol: PRICKLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553036.2 | XP_005553093.1 | 6 - 7 | 860 | 91 | |
| XM_005553037.2 | XP_005553094.1 | 6 - 7 | 718 | 91 | ||
| XM_015449409.1 | XP_015304895.1 | 5 - 6 | 1280 | 91 | ||
| XM_015449410.1 | XP_015304896.1 | 6 - 7 | 860 | 91 | ||
| XM_015449411.1 | XP_015304897.1 | 5 - 6 | 1280 | 91 | ||
| XM_015449412.1 | XP_015304898.1 | 6 - 7 | 1344 | 91 | ||
More Information
Set Membership: |
|

PRICKLE4
Genomic Map