Assay Details
| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593541.2 | XP_005593598.1 | 4 - 5 | 519 | 107 | |
| XM_005593543.2 | XP_005593600.1 | 4 - 5 | 1254 | 107 | ||
More Information
Set Membership: |
|

PRICKLE3
Genomic Map