Assay Details
| Gene Symbol: MGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559250.2 | XP_005559307.1 | 3 - 4 | 2211 | 68 | |
| XM_005559253.2 | XP_005559310.1 | 3 - 4 | 2211 | 68 | ||
| XM_005559254.2 | XP_005559311.1 | 3 - 4 | 2211 | 68 | ||
| XM_005559255.2 | XP_005559312.1 | 3 - 4 | 2211 | 68 | ||
| XM_005559256.2 | XP_005559313.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452686.1 | XP_015308172.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452687.1 | XP_015308173.1 | 3 - 4 | 2173 | 68 | ||
| XM_015452688.1 | XP_015308174.1 | 3 - 4 | 2180 | 68 | ||
| XM_015452689.1 | XP_015308175.1 | 3 - 4 | 2552 | 68 | ||
| XM_015452690.1 | XP_015308176.1 | 3 - 4 | 2179 | 68 | ||
| XM_015452691.1 | XP_015308177.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452692.1 | XP_015308178.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452693.1 | XP_015308179.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452694.1 | XP_015308180.1 | 3 - 4 | 2211 | 68 | ||
| XM_015452695.1 | XP_015308181.1 | 2 - 3 | 1038 | 68 | ||
| XM_015452696.1 | XP_015308182.1 | 2 - 3 | 972 | 68 | ||
More Information
Set Membership: |
|

MGA
Genomic Map