Assay Details
| Gene Symbol: TAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005593235.2 | XP_005593292.1 | 8 - 9 | 2455 | 126 | |
| XM_005593236.2 | XP_005593293.1 | 5 - 6 | 1920 | 126 | ||
| XM_005593238.2 | XP_005593295.1 | 8 - 9 | 2455 | 126 | ||
| XM_015443549.1 | XP_015299035.1 | 7 - 8 | 4021 | 126 | ||
| XM_015443550.1 | XP_015299036.1 | 9 - 10 | 2570 | 126 | ||
| XM_015443551.1 | XP_015299037.1 | 7 - 8 | 2319 | 126 | ||
More Information
Set Membership: |
|

TAB3
Genomic Map