Assay Details
| Gene Symbol: PRKAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283525.1 | NP_001270454.1 | 11 - 12 | 1002 | 73 | |
| XM_005570739.1 | XP_005570796.1 | 11 - 12 | 989 | 73 | ||
| XM_005570740.2 | XP_005570797.1 | 10 - 11 | 1552 | 73 | ||
| XM_005570742.2 | XP_005570799.1 | 10 - 11 | 865 | 73 | ||
| XM_015430614.1 | XP_015286100.1 | 12 - 13 | 1053 | 73 | ||
| GenBank mRNA | AB169499.1 | - | 11 - 12 | 1002 | 73 | |
More Information
Set Membership: |
|

PRKAG1
Genomic Map