Assay Details
| Gene Symbol: ERI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591408.2 | XP_005591465.1 | 5 - 6 | 796 | 128 | |
| XM_005591409.2 | XP_005591466.1 | 6 - 7 | 623 | 128 | ||
| XM_005591410.2 | XP_005591467.1 | 6 - 7 | 580 | 128 | ||
| XM_005591411.2 | XP_005591468.1 | 6 - 7 | 623 | 128 | ||
| XM_015442262.1 | XP_015297748.1 | 6 - 7 | 914 | 128 | ||
| XM_015442264.1 | XP_015297750.1 | 6 - 7 | 917 | 128 | ||
More Information
Set Membership: |
|

ERI2
Genomic Map