Assay Details
| Gene Symbol: DHCR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287695.1 | NP_001274624.1 | 3 - 4 | 564 | 58 | |
| GenBank mRNA | AB125202.1 | - | 3 - 4 | 563 | 58 | |
| AB170728.1 | - | 3 - 4 | 564 | 58 | ||
More Information
Set Membership: |
|

DHCR24
Genomic Map