Assay Details
| Gene Symbol: GTF2H1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284665.1 | NP_001271594.1 | 13 - 14 | 1669 | 84 | |
| XM_015434115.1 | XP_015289601.1 | 13 - 14 | 1954 | 84 | ||
| XM_015434116.1 | XP_015289602.1 | 14 - 15 | 2025 | 84 | ||
| XM_015434117.1 | XP_015289603.1 | 13 - 14 | 1669 | 84 | ||
| XM_015434118.1 | XP_015289604.1 | 13 - 14 | 1701 | 84 | ||
| GenBank mRNA | AB179428.1 | - | 13 - 14 | 1669 | 84 | |
More Information
Set Membership: |
|

GTF2H1
Genomic Map