Assay Details
| Gene Symbol: RFX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559627.2 | XP_005559684.1 | 10 | 7376 | 113 | |
| XM_005559628.2 | XP_005559685.1 | 10 | 7379 | 113 | ||
| XM_015452868.1 | XP_015308354.1 | 9 | 7547 | 113 | ||
More Information
Set Membership: |
|

RFX7
Genomic Map