Assay Details
| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548118.2 | XP_005548175.1 | 2 - 3 | 216 | 101 | |
| XM_015446404.1 | XP_015301890.1 | 2 - 3 | 118 | 101 | ||
More Information
Set Membership: |
|

ATP6V1A
Genomic Map