Assay Details
| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543864.2 | XP_005543921.1 | 5 - 6 | 846 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NT5C1A| Gene Symbol: NT5C1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543864.2 | XP_005543921.1 | 5 - 6 | 846 | 60 | |
Set Membership: |
|