Assay Details
| Gene Symbol: STX7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283441.1 | NP_001270370.1 | 4 - 5 | 442 | 104 | |
| GenBank mRNA | AB173984.1 | - | 4 - 5 | 442 | 104 | |
More Information
Set Membership: |
|

STX7
Genomic Map