Assay Details
| Gene Symbol: ILK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578776.2 | XP_005578833.1 | 6 - 7 | 689 | 103 | |
| XM_005578778.2 | XP_005578835.1 | 5 - 6 | 462 | 103 | ||
| XM_005578779.2 | XP_005578836.1 | 5 - 6 | 523 | 103 | ||
| XM_015435198.1 | XP_015290684.1 | 5 - 6 | 779 | 103 | ||
| XM_015435199.1 | XP_015290685.1 | 5 - 6 | 776 | 103 | ||
More Information
Set Membership: |
|

ILK
Genomic Map