Assay Details
| Gene Symbol: ATPAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543470.2 | XP_005543527.1 | 4 - 5 | 1640 | 129 | |
| GenBank mRNA | AB171005.1 | - | 4 - 5 | 509 | 129 | |
More Information
Set Membership: |
|

ATPAF1
Genomic Map