Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578275.2 | XP_005578332.1 | 1 - 2 | 856 | 55 | |
| XM_005578276.2 | XP_005578333.1 | 1 - 2 | 856 | 55 | ||
| XM_005578277.2 | XP_005578334.1 | 1 - 2 | 856 | 55 | ||
More Information
Set Membership: |
|

WT1
Genomic Map