Assay Details
| Gene Symbol: COPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284810.1 | NP_001271739.1 | 11 - 12 | 1311 | 78 | |
| XM_015437365.1 | XP_015292851.1 | 10 - 11 | 1058 | 78 | ||
| XM_015437367.1 | XP_015292853.1 | 11 - 12 | 1305 | 78 | ||
| XM_015437368.1 | XP_015292854.1 | 9 - 10 | 1227 | 78 | ||
| GenBank mRNA | AB168559.1 | - | 11 - 12 | 1326 | 78 | |
| AB170094.1 | - | 11 - 12 | 1311 | 78 | ||
More Information
Set Membership: |
|

COPS3
Genomic Map