Assay Details
| Gene Symbol: PPP4R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575827.2 | XP_005575884.1 | 3 - 4 | 642 | 63 | |
| XM_005575829.2 | XP_005575886.1 | 3 - 4 | 779 | 63 | ||
| XM_005575830.2 | XP_005575887.1 | 3 - 4 | 779 | 63 | ||
| XM_005575831.2 | XP_005575888.1 | 3 - 4 | 779 | 63 | ||
| XM_005575832.2 | XP_005575889.1 | 3 - 4 | 779 | 63 | ||
| XM_015433645.1 | XP_015289131.1 | 4 - 5 | 1832 | 63 | ||
| XM_015433646.1 | XP_015289132.1 | 3 - 4 | 779 | 63 | ||
| XM_015433647.1 | XP_015289133.1 | 3 - 4 | 779 | 63 | ||
| XM_015433649.1 | XP_015289135.1 | 3 - 4 | 779 | 63 | ||
| XM_015433650.1 | XP_015289136.1 | 3 - 4 | 779 | 63 | ||
| XM_015433651.1 | XP_015289137.1 | 3 - 4 | 779 | 63 | ||
More Information
Set Membership: |
|

PPP4R3B
Genomic Map