Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319524.1 | NP_001306453.1 | 12 - 13 | 1051 | 75 | |
| XM_005582927.2 | XP_005582984.1 | 11 - 12 | 1177 | 75 | ||
| XM_005582928.2 | XP_005582985.1 | 10 - 11 | 1143 | 75 | ||
| XM_015437651.1 | XP_015293137.1 | 9 - 10 | 1024 | 75 | ||
| GenBank mRNA | AB173348.1 | - | 12 - 13 | 1051 | 75 | |
| AB173648.1 | - | 12 - 13 | 1051 | 75 | ||
More Information
Set Membership: |
|

MAP2K4
Genomic Map