Assay Details
| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576409.2 | XP_005576466.1 | 4 - 5 | 587 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC27| Gene Symbol: DNAJC27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576409.2 | XP_005576466.1 | 4 - 5 | 587 | 103 | |
Set Membership: |
|