Assay Details
| Gene Symbol: MMP17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572645.2 | XP_005572702.1 | 7 - 8 | 998 | 116 | |
| XM_015431837.1 | XP_015287323.1 | 6 - 7 | 876 | 116 | ||
More Information
Set Membership: |
|

MMP17
Genomic Map