Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544368.2 | XP_005544425.1 | 1 - 2 | 98 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005544368.2 transcripts.
Set Membership: |
|

PDIK1L
Genomic Map