Assay Details
| Gene Symbol: B3GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540290.2 | XP_005540347.1 | 1 - 2 | 632 | 165 | |
| GenBank mRNA | AB171551.1 | - | 1 - 2 | 439 | 165 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005540290.2 transcripts.
Set Membership: |
|

B3GALT2
Genomic Map