Assay Details
| Gene Symbol: PBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539835.2 | XP_005539892.1 | 4 - 5 | 1004 | 62 | |
| XM_005539837.2 | XP_005539894.1 | 4 - 5 | 1004 | 62 | ||
| XM_015444404.1 | XP_015299890.1 | 3 - 4 | 1012 | 62 | ||
| GenBank mRNA | AB064996.1 | - | 4 - 5 | 832 | 62 | |
More Information
Set Membership: |
|

PBX1
Genomic Map