Assay Details
| Gene Symbol: PTGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285069.1 | NP_001271998.1 | 1 - 2 | 201 | 57 | |
| GenBank mRNA | AB169513.1 | - | 1 - 2 | 201 | 57 | |
| DQ093857.1 | - | 1 - 2 | 94 | 57 | ||
More Information
Set Membership: |
|

PTGDS
Genomic Map