Assay Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283272.1 | NP_001270201.1 | 7 - 8 | 1182 | 81 | |
| GenBank mRNA | AB046046.2 | - | 7 - 8 | 1182 | 81 | |
More Information
Set Membership: |
|

CA10
Genomic Map