Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283904.1 | NP_001270833.1 | 7 - 8 | 1084 | 97 | |
| GenBank mRNA | AB169177.1 | - | 7 - 8 | 1084 | 97 | |
| AB171040.1 | - | 7 - 8 | 916 | 97 | ||
| AB171225.1 | - | 7 - 8 | 1086 | 97 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map