Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578644.2 | XP_005578701.1 | 4 - 5 | 1155 | 92 | |
| XM_015435127.1 | XP_015290613.1 | 3 - 4 | 453 | 92 | ||
| XM_015435128.1 | XP_015290614.1 | 3 - 4 | 607 | 92 | ||
| GenBank mRNA | AB070176.1 | - | 3 - 4 | 828 | 92 | |
More Information
Set Membership: |
|

WEE1
Genomic Map