Assay Details
| Gene Symbol: EIF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005568814.2 | XP_005568871.1 | 4 - 5 | 619 | 73 | |
| XM_005568815.2 | XP_005568872.1 | 4 - 5 | 619 | 73 | ||
| XM_005568816.2 | XP_005568873.1 | 4 - 5 | 426 | 73 | ||
| XM_015457978.1 | XP_015313464.1 | 4 - 5 | 426 | 73 | ||
| XM_015457979.1 | XP_015313465.1 | 4 - 5 | 388 | 73 | ||
| XM_015457980.1 | XP_015313466.1 | 4 - 5 | 584 | 73 | ||
| XM_015457981.1 | XP_015313467.1 | 3 - 4 | 378 | 73 | ||
| XM_015457982.1 | XP_015313468.1 | 3 - 4 | 378 | 73 | ||
More Information
Set Membership: |
|

EIF6
Genomic Map