Assay Details
| Gene Symbol: AK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544063.2 | XP_005544120.1 | 4 - 5 | 578 | 65 | |
| XM_005544065.2 | XP_005544122.1 | 4 - 5 | 509 | 65 | ||
| XM_015438891.1 | XP_015294377.1 | 5 - 6 | 622 | 65 | ||
More Information
Set Membership: |
|

AK2
Genomic Map