Assay Details
| Gene Symbol: PDE6H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570235.2 | XP_005570292.1 | 3 - 4 | 479 | 101 | |
| XM_005570239.2 | XP_005570296.1 | 3 - 4 | 1016 | 101 | ||
| XM_015430420.1 | XP_015285906.1 | 4 - 5 | 641 | 101 | ||
| XM_015430421.1 | XP_015285907.1 | 4 - 5 | 691 | 101 | ||
| XM_015430422.1 | XP_015285908.1 | 3 - 4 | 1274 | 101 | ||
More Information
Set Membership: |
|

PDE6H
Genomic Map