Assay Details
| Gene Symbol: HERPUD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592002.2 | XP_005592059.1 | 6 - 7 | 1378 | 88 | |
| XM_005592004.2 | XP_005592061.1 | 5 - 6 | 1303 | 88 | ||
More Information
Set Membership: |
|

HERPUD1
Genomic Map