Assay Details
| Gene Symbol: IL13RA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594443.2 | XP_005594500.1 | 4 - 5 | 690 | 116 | |
| GenBank mRNA | AY312267.1 | - | 4 - 5 | 439 | 116 | |
More Information
Set Membership: |
|

IL13RA1
Genomic Map