Assay Details
| Gene Symbol: LONRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454455.1 | XP_015309941.1 | 10 - 11 | 1677 | 102 | |
| XM_015454456.1 | XP_015309942.1 | 8 - 9 | 1302 | 102 | ||
| GenBank mRNA | AB168767.1 | - | 9 - 10 | 1428 | 102 | |
| AB174164.1 | - | 2 - 3 | 215 | 102 | ||
More Information
Set Membership: |
|

LONRF1
Genomic Map