Assay Details
| Gene Symbol: CPEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005558550.2 | XP_005558607.1 | 1 - 2 | 2568 | 67 | |
| XM_005558551.2 | XP_005558608.1 | 1 - 2 | 2568 | 67 | ||
| XM_005558552.2 | XP_005558609.1 | 1 - 2 | 2568 | 67 | ||
| XM_005558553.2 | XP_005558610.1 | 1 - 2 | 2568 | 67 | ||
More Information
Set Membership: |
|

CPEB4
Genomic Map