Assay Details
| Gene Symbol: RBFOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591178.2 | XP_005591235.2 | 16 - 17 | 1559 | 65 | |
| XM_005591195.2 | XP_005591252.1 | 10 - 11 | 4810 | 65 | ||
| XM_005591196.2 | XP_005591253.1 | 10 - 11 | 4810 | 65 | ||
| XM_005591198.2 | XP_005591255.1 | 10 - 11 | 4810 | 65 | ||
| XM_015442594.1 | XP_015298080.1 | 15 - 16 | 1520 | 65 | ||
| XM_015442595.1 | XP_015298081.1 | 16 - 17 | 1559 | 65 | ||
| XM_015442597.1 | XP_015298083.1 | 15 - 16 | 1478 | 65 | ||
| XM_015442598.1 | XP_015298084.1 | 15 - 16 | 1466 | 65 | ||
| XM_015442599.1 | XP_015298085.1 | 16 - 17 | 1559 | 65 | ||
| XM_015442601.1 | XP_015298087.1 | 16 - 17 | 1559 | 65 | ||
More Information
Set Membership: |
|

RBFOX1
Genomic Map