Assay Details
| Gene Symbol: LOC102123567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559572.2 | XP_005559629.1 | 8 - 9 | 1684 | 63 | |
| XM_015452832.1 | XP_015308318.1 | 8 - 9 | 1682 | 63 | ||
| GenBank mRNA | AB172691.1 | - | 9 - 10 | 1214 | 63 | |
More Information
Set Membership: |
|

LOC102123567
Genomic Map