Assay Details
| Gene Symbol: SPAG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283739.1 | NP_001270668.1 | 4 - 5 | 343 | 70 | |
| GenBank mRNA | AB172634.1 | - | 4 - 5 | 343 | 70 | |
More Information
Set Membership: |
|

SPAG7
Genomic Map