Assay Details
| Gene Symbol: LOC102121477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550998.2 | XP_005551055.2 | 10 - 11 | 1371 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102121477| Gene Symbol: LOC102121477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550998.2 | XP_005551055.2 | 10 - 11 | 1371 | 79 | |
Set Membership: |
|