Assay Details
| Gene Symbol: PLEKHG4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015451190.1 | XP_015306676.1 | 1 - 2 | 400 | 76 | |
| XM_015451191.1 | XP_015306677.1 | 1 - 2 | 400 | 76 | ||
| XM_015451192.1 | XP_015306678.1 | 1 - 2 | 400 | 76 | ||
| XM_015451193.1 | XP_015306679.1 | 1 - 2 | 400 | 76 | ||
| XM_015451194.1 | XP_015306680.1 | 1 - 2 | 400 | 76 | ||
| XM_015451196.1 | XP_015306682.1 | 1 - 2 | 400 | 76 | ||
More Information
Set Membership: |
|

PLEKHG4B
Genomic Map