Assay Details
| Gene Symbol: MFSD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576446.2 | XP_005576503.1 | 11 - 12 | 2006 | 68 | |
| XM_015433947.1 | XP_015289433.1 | 9 - 10 | 1541 | 68 | ||
More Information
Set Membership: |
|

MFSD2B
Genomic Map