Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589158.2 | XP_005589215.1 | 6 - 7 | 950 | 90 | |
| XM_005589159.2 | XP_005589216.1 | 5 - 6 | 1151 | 90 | ||
| XM_015442049.1 | XP_015297535.1 | 6 - 7 | 1052 | 90 | ||
| XM_015442050.1 | XP_015297536.1 | 5 - 6 | 1498 | 90 | ||
More Information
Set Membership: |
|

ACP7
Genomic Map