Assay Details
| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005591035.2 | XP_005591092.1 | 5 - 6 | 1394 | 59 | |
| XM_005591037.2 | XP_005591094.1 | 2 - 3 | 389 | 59 | ||
| XM_015442613.1 | XP_015298099.1 | 5 - 6 | 1233 | 59 | ||
More Information
Set Membership: |
|

BICDL2
Genomic Map