Assay Details
| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541223.2 | XP_005541280.1 | 7 - 8 | 1569 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NECTIN4| Gene Symbol: NECTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541223.2 | XP_005541280.1 | 7 - 8 | 1569 | 76 | |
Set Membership: |
|